trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-4-((4-(trans-4-propylcyclohexyl)phenyl)ethynyl)benzene

A145519-1G A145519-250MG A145519-25G
76811-960EA 14.86 USD
76811-960 76811-962 76811-964
trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene 97%
trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₂₅H₃₀
MW: 330.515 g/mol
MDL Number: MFCD11053405
CAS Number: 107949-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR