(4-(Hydroxymethyl)-2-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Hydroxymethyl)-2-methylphenylboronic acid

A144652-100MG A144652-1G A144652-250MG
76811-910EA 35 USD
76811-910 76811-912 76811-914
(4-(Hydroxymethyl)-2-methylphenyl)boronic acid 98%
(4-(Hydroxymethyl)-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD13181612
CAS Number: 166386-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR