7H-Purin-6-amine phosphate 99%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopurine phosphate Vitamin B4, Adenine phosphate

A144524-100G A144524-25G A144524-500G
76811-904EA 58.35 USD
76811-904 76811-906 76811-908
7H-Purin-6-amine phosphate 99%
7H-Purin-6-amine phosphate

Precaución: for Research Use Only

Formula: C₅H₈N₅O₄P
MW: 233.12 g/mol
MDL Number: MFCD00035170
CAS Number: 70700-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR