2-(Bromomethyl)-6-fluoropyridine 95%

Supplier: Ambeed
Danger

A144299-100MG A144299-1G A144299-250MG
76811-894EA 15 USD
76811-894 76811-896 76811-898
2-(Bromomethyl)-6-fluoropyridine 95%
2-(Bromomethyl)-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MDL Number: MFCD09953532
CAS Number: 100202-78-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR