5-Bromo-2-methoxyisonicotinaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxypyridine-4-carboxaldehyde

A143705-100MG A143705-1G A143705-250MG A143705-5G A143705-25G
76811-876EA 7.43 USD
76811-876 76811-878 76811-880 76811-882 77566-882
5-Bromo-2-methoxyisonicotinaldehyde 98%
5-Bromo-2-methoxyisonicotinaldehyde

Precaución: For research use only.

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD11856042
CAS Number: 936011-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR