N-Hydroxy-4-methylbenzenesulfonamide 98%

Supplier: Ambeed
Danger

A143700-1G A143700-250MG A143700-25G A143700-5G
76811-868EA 76 USD
76811-868 76811-870 76811-872 76811-874
N-Hydroxy-4-methylbenzenesulfonamide 98%
N-Hydroxy-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₇H₉NO₃S
MDL Number: MFCD11850835
CAS Number: 1593-60-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR