(R)-Morpholin-2-ylmethanol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A143087-100MG
A143087-250MG
A143087-5G
A143087-1G
76811-852EA
11
USD
76811-852
76811-854
77566-870
77671-042
(R)-Morpholin-2-ylmethanol hydrochloride 95%
(R)-Morpholin-2-ylmethanol hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₁NO₂·HCl |
MDL Number:
MFCD11052541 CAS Number: 1436436-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...