5-Bromo-4-methyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

A142975-100MG A142975-1G A142975-250MG
76811-846EA 14 USD
76811-846 76811-848 76811-850
5-Bromo-4-methyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-4-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MDL Number: MFCD12024533
CAS Number: 1150617-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR