Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A142271-100MG
A142271-1G
A142271-250MG
76811-812EA
25.47
USD
76811-812
76811-814
76811-816
Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate 95%
Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₇N₃O₂S MW: 209.225 g/mol |
MDL Number:
MFCD00204059 CAS Number: 244014-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...