2-Bromo-3,5-dichloropyrazine 97%

Supplier: Ambeed
Warning

A141928-1G A141928-250MG
76811-806EA 31.83 USD
76811-806 76811-808
2-Bromo-3,5-dichloropyrazine 97%
2-Bromo-3,5-dichloropyrazine

Precaución: for Research Use Only

Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
MDL Number: MFCD14581976
CAS Number: 1206249-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR