4-Bromo-o-toluidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-bromotoluene, 4-Bromo-2-methylaniline
A141877-1000G
A141877-100G
A141877-10G
A141877-1G
A141877-500G
A141877-5G
A141877-25G
76811-792EA
269.47
USD
76811-792
76811-794
76811-796
76811-798
76811-800
76811-802
77316-262
4-Bromo-o-toluidine 98%
4-Bromo-o-toluidine
Precaución: for Research Use Only
Formula:
C₇H₈BrN MW: 186.05 g/mol |
MDL Number:
MFCD00007825 CAS Number: 583-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...