3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium 95%

Supplier: Ambeed
Warning

Synonyms: Nicotinamide riboside

A141741-1G A141741-5G A141741-25G
76811-790EA 8.49 USD
76811-790 77566-854 77611-294
3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium 95%
3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium

Precaución: For research use only.

Formula: C₁₁H₁₅N₂O₅
MW: 255.24 g/mol
MDL Number: MFCD16621007
CAS Number: 1341-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR