trans-4-Aminotetrahydrofuran-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

A141244-100MG A141244-1G A141244-250MG A141244-5G
76811-782EA 54 USD
76811-782 76811-784 76811-786 76811-788
trans-4-Aminotetrahydrofuran-3-ol hydrochloride 97%
trans-4-Aminotetrahydrofuran-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉NO₂·ClH
MDL Number: MFCD11520524
CAS Number: 215940-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR