5-Bromo-1H-1,2,4-triazol-3-amine 95%

Supplier: Ambeed
Warning

A140898-100MG A140898-1G A140898-5G A140898-250MG A140898-25G
76811-774EA 13 USD
76811-774 77671-008 77671-010 77883-126 77883-127
5-Bromo-1H-1,2,4-triazol-3-amine 95%
5-Bromo-1H-1,2,4-triazol-3-amine

Precaución: for Research Use Only

Formula: C₂H₃BrN₄
MDL Number: MFCD18807020
CAS Number: 389122-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR