2-Bromo-6,7-dihydrothieno[3,2-c]pyridin-4(5H)-one 97%

Supplier: Ambeed
Warning

A140315-100MG A140315-1G A140315-250MG
76811-736EA 58.35 USD
76811-736 77611-278 77671-002
2-Bromo-6,7-dihydrothieno[3,2-c]pyridin-4(5H)-one 97%
2-Bromo-6,7-dihydrothieno[3,2-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₆BrNOS
MW: 232.1 g/mol
MDL Number: MFCD11656583
CAS Number: 1078150-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR