2,4-Difluoro-3-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A139985-1G A139985-250MG
76811-728EA 87 USD
76811-728 76811-730
2,4-Difluoro-3-hydroxybenzaldehyde 95%
2,4-Difluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MW: 158.1000 g/mol
MDL Number: MFCD12405741
CAS Number: 192927-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR