2-Amino-5-nitropyridin-3-ol 95%

Supplier: Ambeed
Warning

A139761-10G A139761-1G A139761-250MG A139761-5G
76811-718EA 420 USD
76811-718 76811-720 76811-722 77883-120
2-Amino-5-nitropyridin-3-ol 95%
2-Amino-5-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₅N₃O₃
MW: 155.11 g/mol
MDL Number: MFCD13193356
CAS Number: 908248-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR