(1-Aminocyclohexyl)acetic acid 95%

Supplier: Ambeed
Warning

A139497-100MG A139497-1G A139497-250MG A139497-5G
76811-706EA 8 USD
76811-706 76811-708 76811-710 76811-712
(1-Aminocyclohexyl)acetic acid 95%
(1-Aminocyclohexyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD02258116
CAS Number: 37631-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR