3-Methylthiophene-2-carbonyl chloride 98%

Supplier: Ambeed
Danger

A139217-1G A139217-5G
76811-676EA 23.34 USD
76811-676 76811-678
3-Methylthiophene-2-carbonyl chloride 98%
3-Methylthiophene-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C₆H₅ClOS
MW: 160.62 g/mol
MDL Number: MFCD00130091
CAS Number: 61341-26-2
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR