1-Cyclohexene-1-carboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: Cyclohex-1-enecarboxylic acid

A138536-1G A138536-25G A138536-5G A138536-10G
76811-648EA 14.86 USD
76811-648 76811-650 76811-652 77566-786
1-Cyclohexene-1-carboxylic acid 97%
1-Cyclohexene-1-carboxylic acid

Precaución: For research use only.

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD00001545
CAS Number: 636-82-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR