3,5-Dimethyl-4-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A138431-10G A138431-1G A138431-250MG A138431-5G
76811-642EA 62 USD
76811-642 76811-644 76811-646 77670-980
3,5-Dimethyl-4-fluorobenzonitrile 98%
3,5-Dimethyl-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈FN
MDL Number: MFCD09832248
CAS Number: 867367-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR