3-Amino-6-chloropyrazine-2-carboxamide 95%

Supplier: Ambeed
Warning

A137602-100MG A137602-1G A137602-250MG
76811-606EA 27.59 USD
76811-606 76811-608 76811-610
3-Amino-6-chloropyrazine-2-carboxamide 95%
3-Amino-6-chloropyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₅ClN₄O
MW: 172.57 g/mol
MDL Number: MFCD05722610
CAS Number: 1125-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR