4-Amino-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A137426-10G A137426-1G A137426-25G A137426-5G
76811-586EA 37 USD
76811-586 76811-588 76811-590 76811-592
4-Amino-2-nitrobenzoic acid 98%
4-Amino-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MDL Number: MFCD00189376
CAS Number: 610-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR