6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 96%

Supplier: Ambeed
Warning

A137395-1G A137395-250MG A137395-5G
76811-580EA 72.15 USD
76811-580 76811-582 76811-584
6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 96%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
MDL Number: MFCD10697640
CAS Number: 383875-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR