Pyrimidin-4-ol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxypyrimidine

A136378-100G A136378-10G A136378-1G A136378-25G A136378-5G
76811-530EA 198.39 USD
76811-530 76811-532 76811-534 76811-536 76811-538
Pyrimidin-4-ol 97%
Pyrimidin-4-ol

Precaución: for Research Use Only

Formula: C4H4N2O
MW: 96.0874 g/mol
MDL Number: MFCD00038024
CAS Number: 51953-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR