(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-fluoro-3-propoxyphenylboronic acid

A136288-1G A136288-25G A136288-5G
76811-516EA 26 USD
76811-516 76811-518 76811-520
(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid 95%
(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BBrFO₃
MDL Number: MFCD09038422
CAS Number: 1072951-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR