7-Bromo-4,5-dihydro-1H-benzo[b][1,4]diazepin-2(3H)-one 95%

Supplier: Ambeed
Warning

A136234-100MG A136234-250MG A136234-50MG
76811-510EA 80 USD
76811-510 76811-512 76811-514
7-Bromo-4,5-dihydro-1H-benzo[b][1,4]diazepin-2(3H)-one 95%
7-Bromo-4,5-dihydro-1H-benzo[b][1,4]diazepin-2(3H)-one

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MW: 241.08 g/mol
MDL Number: MFCD11101049
CAS Number: 219686-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR