4-Methyl-2H-1,2,3-triazole 97%

Supplier: Ambeed
Warning

A136187-100MG A136187-250MG A136187-1G
76811-506EA 54 USD
76811-506 76811-508 77883-100
4-Methyl-2H-1,2,3-triazole 97%
4-Methyl-2H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₅N₃
MDL Number: MFCD20482150
CAS Number: 19230-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR