Cinnamic acid 98% (only trans isomers)

Supplier: Ambeed
Warning

A135758-100G A135758-500G A135758-1000G A135758-1G
76811-494EA 8.49 USD
76811-494 76811-496 77566-676 77566-678
Cinnamic acid 98% (only trans isomers)
Cinnamic acid

Precaución: For research use only.

Formula: C₉H₈O₂
MW: 148.15 g/mol
MDL Number: MFCD00004369
CAS Number: 621-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR