Ethyl 2,6-dichloro-4-methylnicotinate 98%

Supplier: Ambeed
Warning

A135165-1G A135165-250MG
76811-476EA 76 USD
76811-476 76811-478
Ethyl 2,6-dichloro-4-methylnicotinate 98%
Ethyl 2,6-dichloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD21604262
CAS Number: 108130-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR