5-Fluoro-2-(hydroxymethyl)benzeneboronic acid 97%

Supplier: Ambeed
Warning

A134905-250MG A134905-1G A134905-5G
76811-470EA 19 USD
76811-470 77883-091 77883-092
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid 97%
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MDL Number: MFCD11111910
CAS Number: 1246633-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR