Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Pyr-OBzl

A134826-100G A134826-10G A134826-25G A134826-500G
76811-448EA 17 USD
76811-448 76811-450 76811-452 76811-454 76811-456
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 97%
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₅
MDL Number: MFCD04115781
CAS Number: 113400-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR