5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A134490-100MG A134490-1G A134490-5G
76811-404EA 30.77 USD
76811-404 76811-406 76811-408
5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 95%
5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD12962791
CAS Number: 1190314-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR