tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl azetidin-3-yl(methyl)carbamate hydrochloride

A133986-100MG A133986-10G A133986-1G A133986-250MG A133986-25G A133986-5G
76811-380EA 5 USD
76811-380 76811-382 76811-384 76811-386 76811-388 76811-390
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride 95%
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD12963843
CAS Number: 943060-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR