(S)-(+)-Thiazolidine-4-carboxylic acid 95%

Supplier: Ambeed
Danger

Synonyms: H-D-Thz-OH, (S)-Thiazolidine-4-carboxylic acid, D-Thioproline

A133716-10G A133716-250MG A133716-5G A133716-1G A133716-25G
76811-364EA 113.52 USD
76811-364 76811-366 76811-368 77670-908 77670-910
(S)-(+)-Thiazolidine-4-carboxylic acid 95%
(S)-(+)-Thiazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₇NO₂S
MW: 133.17 g/mol
MDL Number: MFCD00137425
CAS Number: 45521-09-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR