1-Boc-3-acetyl-piperidine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-acetylpiperidine-1-carboxylate

A133687-100MG A133687-10G A133687-1G A133687-250MG A133687-5G
76811-350EA 21.22 USD
76811-350 76811-352 76811-354 76811-356 76811-358
1-Boc-3-acetyl-piperidine 95%
1-Boc-3-acetyl-piperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD09953372
CAS Number: 858643-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR