(S)-tert-Butyl (5-oxopyrrolidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A133557-100MG A133557-250MG
76811-342EA 65.78 USD
76811-342 77670-902
(S)-tert-Butyl (5-oxopyrrolidin-3-yl)carbamate 95%
(S)-tert-Butyl (5-oxopyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
MW: 200.24 g/mol
MDL Number: MFCD20257110
CAS Number: 672883-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR