3,6-Dimethoxypyridazine 97%

Supplier: Ambeed
Warning

A133382-100MG A133382-10G A133382-1G A133382-250MG A133382-25G
76811-334EA 5 USD
76811-334 76811-336 76811-338 76811-340 77883-076
3,6-Dimethoxypyridazine 97%
3,6-Dimethoxypyridazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD00234224
CAS Number: 4603-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR