(2-Chloro-6-nitrophenyl)methanamine 95%

Supplier: Ambeed
Warning

A132995-100MG A132995-1G A132995-250MG
76811-322EA 93.36 USD
76811-322 76811-324 76811-326
(2-Chloro-6-nitrophenyl)methanamine 95%
(2-Chloro-6-nitrophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
MDL Number: MFCD03410976
CAS Number: 101252-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR