2-Phenylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-1H-imidazole

A132487-100G A132487-10G A132487-25G A132487-500G
76811-294EA 12.74 USD
76811-294 76811-296 77314-322 77611-180
2-Phenylimidazole 98%
2-Phenylimidazole

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
MDL Number: MFCD00005186
CAS Number: 670-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR