4-(2-Chlorophenoxy)-N-(3-(methylcarbamoyl)phenyl)piperidine-1-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: A939572

A132282-10MG A132282-1MG A132282-2MG A132282-50MG A132282-5MG A132282-100MG A132282-250MG
76811-282EA 42.44 USD
76811-282 76811-284 76811-286 76811-288 76811-290 77670-876 77670-878
4-(2-Chlorophenoxy)-N-(3-(methylcarbamoyl)phenyl)piperidine-1-carboxamide 98%
4-(2-Chlorophenoxy)-N-(3-(methylcarbamoyl)phenyl)piperidine-1-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₂₂ClN₃O₃
MW: 387.86 g/mol
MDL Number: MFCD10698997
CAS Number: 1032229-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR