(S)-L-Benzyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxyl 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-4-Oxo-Pro-OMe, (S)-1-Benzyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate
A132265-250MG
A132265-5G
A132265-1G
A132265-25G
76811-276EA
36.08
USD
76811-276
76811-278
77611-174
77670-874
(S)-L-Benzyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxyl 95%
(S)-L-Benzyl-2-methyl-4-oxopyrrolidine-1,2-dicarboxyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₅NO₅ MW: 277.28 g/mol |
MDL Number:
MFCD09608116 CAS Number: 16217-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...