3-(Cbz-amino)azetidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl azetidin-3-ylcarbamate hydrochloride

A132219-10G A132219-1G A132219-250MG A132219-25G A132219-5G
76811-262EA 238.71 USD
76811-262 76811-264 76811-266 76811-268 76811-270
3-(Cbz-amino)azetidine hydrochloride 97%
3-(Cbz-amino)azetidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₂·ClH
MW: 242.702 g/mol
MDL Number: MFCD11101353
CAS Number: 1203099-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR