Ethyl-3-bromo-4-methyl-1H-pyrazole-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A131975-100MG
A131975-1G
A131975-250MG
A131975-5G
A131975-25G
76811-240EA
14
USD
76811-240
76811-242
76811-244
76811-246
77881-045
Ethyl-3-bromo-4-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl-3-bromo-4-methyl-1H-pyrazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉BrN₂O₂ |
MDL Number:
MFCD20921911 CAS Number: 1257861-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...