4,6-Dibromopyridin-2-amine 98%

Supplier: Ambeed
Warning

A131962-100MG A131962-1G A131962-250MG
76811-236EA 75 USD
76811-236 77883-063 77883-064
4,6-Dibromopyridin-2-amine 98%
4,6-Dibromopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MDL Number: MFCD18447569
CAS Number: 408352-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR