2-Bromo-4-fluorophenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Bromo-4-fluorophenyl)acetic acid

A131224-100G A131224-10G A131224-25G A131224-500G A131224-5G
76811-196EA 136.86 USD
76811-196 76811-198 76811-200 76811-202 76811-204
2-Bromo-4-fluorophenylacetic acid 97%
2-Bromo-4-fluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.0300 g/mol
MDL Number: MFCD04107675
CAS Number: 61150-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR