5-Bromo-2-mercaptobenzothiazole 97%

Supplier: Ambeed
Warning

A131083-1G A131083-250MG A131083-5G
76811-190EA 30.77 USD
76811-190 76811-192 77670-850
5-Bromo-2-mercaptobenzothiazole 97%
5-Bromo-2-mercaptobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₄BrNS₂
MDL Number: MFCD08460052
CAS Number: 71216-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR