4-Amino-6-chloropyrimidine 97%

Supplier: Ambeed
Warning

A130724-100G A130724-10G A130724-25G A130724-500G A130724-5G A130724-1000G
76811-162EA 21 USD
76811-162 76811-164 76811-166 76811-168 76811-170 77883-050
4-Amino-6-chloropyrimidine 97%
4-Amino-6-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
MDL Number: MFCD00053576
CAS Number: 5305-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR