4'-(Benzyloxy)benzylidene-4-fluoroaniline 95%

Supplier: Ambeed
Warning

A130033-100G A130033-10G A130033-500G A130033-5G
76811-130EA 90 USD
76811-130 76811-132 76811-134 76811-136 76811-138
4'-(Benzyloxy)benzylidene-4-fluoroaniline 95%
4'-(Benzyloxy)benzylidene-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₂₀H₁₆FNO
MDL Number: MFCD00017951
CAS Number: 70627-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR