(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

A129793-100MG A129793-10G A129793-1G A129793-250MG A129793-25G A129793-5G
76811-116EA 5 USD
76811-116 76811-118 76811-120 76811-122 76811-124 76811-126
(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride 97%
(S)-3-Piperidinecarboxylic acid methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MDL Number: MFCD09271813
CAS Number: 164323-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR